This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363989_g1
See other GPX4 GE Assays ›
Gene Symbol:
GPX4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
-
Chromosome Location:
Chr.28: 3034818 - 3036391 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363989_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001346448.1 NP_001333377.1 5 - 6 499 80
NM_001346449.1 NP_001333378.1 5 - 6 508 80
NM_204220.2 NP_989551.2 5 - 6 508 80
GenBank mRNA AF498316.2 - 5 - 6 453 80
BU422188.1 - 3 - 4 316 80
CD215039.1 - 5 - 6 476 80
CD733308.1 - 1 - 2 33 80
CN217579.1 - 5 - 6 482 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0