This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03364617_m1
See other QKI GE Assays ›
Gene Symbol:
QKI
Gene Name:
QKI, KH domain containing RNA binding
Gene Aliases:
-
Chromosome Location:
Chr.3: 44204197 - 44347294 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03364617_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: QKI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204310.2 NP_989641.2 4 - 5 588 99
XM_015283998.2 XP_015139484.1 5 - 6 1139 99
XM_015283999.2 XP_015139485.1 5 - 6 1141 99
XM_015284000.2 XP_015139486.1 5 - 6 1133 99
XM_015284001.2 XP_015139487.1 5 - 6 1140 99
XM_015284002.2 XP_015139488.1 5 - 6 1140 99
XM_015284003.2 XP_015139489.1 4 - 5 1083 99
XM_015284004.2 XP_015139490.1 5 - 6 1140 99
XM_025148481.1 XP_025004249.1 5 - 6 1140 99
GenBank mRNA U86764.1 - 4 - 5 588 99
U86766.1 - 5 - 6 639 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
regulation of translation
multicellular organism development
RNA splicing
cell differentiation
mRNA transport
mRNA binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0