This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03365365_s1
See other ARC GE Assays ›
Gene Symbol:
ARC
Gene Name:
activity regulated cytoskeleton associated protein
Gene Aliases:
ARG3.1
Chromosome Location:
Chr.2: 147995617 - 148001989 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03365365_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204432.1 NP_989763.1 1 1148 65
XM_015283164.2 XP_015138650.1 1 1793 65
GenBank mRNA AJ272062.1 - 1 1148 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cytoskeleton organization
endoderm development
long-term memory
anterior/posterior pattern specification
cell migration
regulation of cell morphogenesis
regulation of neuronal synaptic plasticity
modulation of synaptic transmission
mRNA transport
protein homooligomerization
regulation of postsynaptic neurotransmitter receptor internalization
vesicle-mediated intercellular transport
regulation of long-term synaptic potentiation
regulation of long term synaptic depression
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0