This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03366567_m1
See other NFE2L2 GE Assays ›
Gene Symbol:
NFE2L2
Gene Name:
nuclear factor, erythroid 2 like 2
Gene Aliases:
NRF2
Chromosome Location:
Chr.7: 15795250 - 15819379 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03366567_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFE2L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205117.1 NP_990448.1 4 - 5 611 68
XM_015289381.2 XP_015144867.1 3 - 4 532 68
XM_015289382.2 XP_015144868.1 4 - 5 696 68
XM_015289383.2 XP_015144869.1 4 - 5 703 68
XM_015289385.2 XP_015144871.1 3 - 4 715 68
XM_015289386.2 XP_015144872.1 3 - 4 594 68
XM_015289387.2 XP_015144873.1 3 - 4 434 68
XM_025152143.1 XP_025007911.1 3 - 4 404 68
XM_025152144.1 XP_025007912.1 4 - 5 616 68
XM_025152145.1 XP_025007913.1 5 - 6 1808 68
XM_025152146.1 XP_025007914.1 3 - 4 425 68
XM_025152148.1 XP_025007916.1 3 - 4 415 68
GenBank mRNA D49365.1 - 4 - 5 611 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline