This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03366968_m1
See other FGF1 GE Assays ›
Gene Symbol:
FGF1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
ECGF, FGF-1
Chromosome Location:
Chr.13: 17962470 - 17981469 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03366968_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205180.1 NP_990511.1 2 - 3 217 107
XM_015294009.2 XP_015149495.1 2 - 3 338 107
XM_015294010.2 XP_015149496.1 3 - 4 407 107
XM_015294011.1 XP_015149497.1 2 - 3 353 107
XM_025154814.1 XP_025010582.1 3 - 4 325 107
XM_025154815.1 XP_025010583.1 2 - 3 258 107
XM_025154816.1 XP_025010584.1 3 - 4 324 107
XM_025154817.1 XP_025010585.1 2 - 3 305 107
GenBank mRNA S63263.1 - 2 - 3 217 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
angiogenesis
organ induction
positive regulation of protein phosphorylation
epicardial cell to mesenchymal cell transition
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
organ morphogenesis
positive regulation of endothelial cell migration
positive regulation of gene expression
cell differentiation
lung development
regulation of cell migration
positive regulation of cell migration
activation of protein kinase B activity
cellular response to heat
wound healing
positive regulation of MAP kinase activity
positive regulation of cholesterol biosynthetic process
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
positive regulation of cell division
branch elongation involved in ureteric bud branching
mesonephric epithelium development
regulation of endothelial tube morphogenesis
positive regulation of intracellular signal transduction
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
fibroblast growth factor receptor binding
integrin binding
growth factor activity
heparin binding
S100 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline