This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03371403_m1
See other CAPZB GE Assays ›
Gene Symbol:
CAPZB
Gene Name:
capping actin protein of muscle Z-line beta subunit
Gene Aliases:
-
Chromosome Location:
Chr.21: 4686546 - 4739397 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03371403_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAPZB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001173529.1 NP_001167000.1 4 - 5 443 68
NM_205437.2 NP_990768.1 4 - 5 443 68
XM_015296835.2 XP_015152321.1 4 - 5 479 68
GenBank mRNA J04959.1 - 3 - 4 356 68
U07826.1 - 4 - 5 388 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytoskeleton organization
actin polymerization or depolymerization
regulation of lamellipodium assembly
regulation of cell morphogenesis
lamellipodium assembly
barbed-end actin filament capping
negative regulation of filopodium assembly
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0