This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03372858_s1
See other HSD3B1 GE Assays ›
Gene Symbol:
HSD3B1
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Gene Aliases:
HSD3B2
Chromosome Location:
Chr.1: 79311683 - 79327343 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03372858_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD3B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205118.1 NP_990449.1 4 1057 125
GenBank mRNA D43762.1 - 4 1057 125
D43763.1 - 3 1049 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0