This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03644103_m1
See other PNPLA4 GE Assays ›
Gene Symbol:
PNPLA4
Gene Name:
patatin like phospholipase domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 127493302 - 127513081 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03644103_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNPLA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130740.1 NP_001124212.1 4 - 5 478 110
XM_015273977.2 XP_015129463.2 7 - 8 1089 110
XM_015273984.2 XP_015129470.2 7 - 8 1113 110
XM_015273987.2 XP_015129473.2 7 - 8 965 110
XM_015273997.2 XP_015129483.2 7 - 8 1088 110
XM_015274000.2 XP_015129486.2 7 - 8 1009 110
XM_025149860.1 XP_025005628.1 4 - 5 388 110
XM_025149863.1 XP_025005631.1 6 - 7 742 110
GenBank mRNA EU419877.1 - 4 - 5 478 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0