This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03644104_m1
See other PNPLA4 GE Assays ›
Gene Symbol:
PNPLA4
Gene Name:
patatin like phospholipase domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 127493302 - 127513081 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03644104_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNPLA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130740.1 NP_001124212.1 5 - 6 625 140
XM_015273977.2 XP_015129463.2 8 - 9 1236 140
XM_015273984.2 XP_015129470.2 8 - 9 1260 140
XM_015273987.2 XP_015129473.2 8 - 9 1112 140
XM_015273997.2 XP_015129483.2 8 - 9 1235 140
XM_015274000.2 XP_015129486.2 8 - 9 1156 140
XM_025149860.1 XP_025005628.1 5 - 6 535 140
XM_025149863.1 XP_025005631.1 7 - 8 889 140
GenBank mRNA EU419877.1 - 5 - 6 625 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0