This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03814404_s1
See other RARA GE Assays ›
Gene Symbol:
RARA
Gene Name:
retinoic acid receptor alpha
Gene Aliases:
RAR-ALPHA1
Chromosome Location:
Chr.27: 7222549 - 7232608 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03814404_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RARA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204536.1 NP_989867.1 5 748 67
GenBank mRNA X73972.1 - 748 67
X78333.1 - 629 67
X78335.1 - 679 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
neural tube closure
glandular epithelial cell development
outflow tract septum morphogenesis
growth plate cartilage development
protein phosphorylation
germ cell development
spermatogenesis
positive regulation of cell proliferation
hormone-mediated signaling pathway
hindbrain morphogenesis
cell differentiation
negative regulation of granulocyte differentiation
embryonic camera-type eye development
response to retinoic acid
negative regulation of interferon-gamma production
negative regulation of tumor necrosis factor production
positive regulation of interleukin-13 production
positive regulation of interleukin-4 production
positive regulation of interleukin-5 production
multicellular organism growth
negative regulation of apoptotic process
apoptotic cell clearance
positive regulation of T-helper 2 cell differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
positive regulation of binding
ventricular cardiac muscle cell differentiation
Sertoli cell fate commitment
limb development
face development
trachea cartilage development
chondroblast differentiation
negative regulation of cartilage development
cellular response to lipopolysaccharide
cellular response to retinoic acid
cellular response to estrogen stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
retinoic acid binding
transcription coactivator activity
transcription corepressor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
receptor binding
zinc ion binding
protein domain specific binding
chromatin DNA binding
protein kinase B binding
retinoic acid-responsive element binding
protein kinase A binding
alpha-actinin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0