This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07155928_m1
See other AWAT1 GE Assays ›
Gene Symbol:
AWAT1
Gene Name:
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 1218114 - 1224786 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07155928_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AWAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318429.1 NP_001305358.1 1 - 2 40 55
XM_004940542.3 XP_004940599.1 1 - 2 999 55
XM_015278337.2 XP_015133823.1 1 - 2 999 55
XM_015278338.2 XP_015133824.1 1 - 2 999 55
XM_015278339.2 XP_015133825.1 1 - 2 999 55
XM_015278340.2 XP_015133826.1 1 - 2 999 55
XM_015278341.2 XP_015133827.1 1 - 2 999 55
XM_015278342.2 XP_015133828.1 1 - 2 999 55
GenBank mRNA JQ031644.1 - 1 - 2 40 55
KU853995.1 - 1 - 2 40 55

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015278340.2, XM_015278341.2, XM_015278342.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0