This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07157297_g1
See other P2RX3 GE Assays ›
Gene Symbol:
P2RX3
Gene Name:
purinergic receptor P2X 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 17278095 - 17282397 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07157297_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001329216.1 NP_001316145.1 1 - 2 183 57
XM_025150827.1 XP_025006595.1 2 - 3 588 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0