This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07157572_m1
See other TNFRSF4 GE Assays ›
Gene Symbol:
TNFRSF4
Gene Name:
TNF receptor superfamily member 4
Gene Aliases:
-
Chromosome Location:
Chr.21: 2547599 - 2557418 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07157572_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001354724.1 NP_001341653.1 5 - 6 673 75
XM_025142474.1 XP_024998242.1 6 - 7 2898 75
XM_025142475.1 XP_024998243.1 6 - 7 2887 75
GenBank mRNA CR354356.1 - 3 - 4 1172 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0