This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07158021_g1
See other GJC3 GE Assays ›
Gene Symbol:
GJC3
Gene Name:
gap junction protein gamma 3
Gene Aliases:
GJA7, GJC1
Chromosome Location:
Chr.27: 3660155 - 3672000 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07158021_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205503.1 NP_990834.1 2 - 3 185 103
XM_015299213.2 XP_015154699.1 3 - 4 437 103
XM_015299219.2 XP_015154705.1 3 - 4 1992 103
XM_015299220.2 XP_015154706.1 3 - 4 322 103
XM_015299221.2 XP_015154707.1 3 - 4 513 103
XM_025143956.1 XP_024999724.1 5 - 6 2688 103
XM_025143957.1 XP_024999725.1 5 - 6 3263 103
GenBank mRNA M35044.1 - 2 - 3 185 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_205503.1, XM_015299213.2, XM_015299219.2, XM_015299220.2, XM_015299221.2, XM_025143956.1, XM_025143957.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
cell-cell signaling
chemical synaptic transmission
visual perception
gap junction assembly
ion transmembrane transport
cell development
cardiac muscle tissue development
AV node cell to bundle of His cell communication by electrical coupling
ion channel activity
gap junction channel activity
gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline