This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07158031_m1
See other RBM12 GE Assays ›
Gene Symbol:
RBM12
Gene Name:
RNA binding motif protein 12
Gene Aliases:
-
Chromosome Location:
Chr.20: 1079716 - 1091358 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07158031_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_206986.2 NP_996869.2 1 - 2 117 81
GenBank mRNA AY239220.1 - 1 - 2 108 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_206986.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0