This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07158682_g1
See other ROMO1 GE Assays ›
Gene Symbol:
ROMO1
Gene Name:
reactive oxygen species modulator 1
Gene Aliases:
C20orf52
Chromosome Location:
Chr.20: 1061506 - 1063498 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07158682_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROMO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198821.1 NP_001185750.1 1 - 2 52 93
XM_015296248.2 XP_015151734.1 1 - 2 64 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001198821.1, XM_015296248.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0