Assay Details
Gene Symbol: TP53I11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257369.3 | NP_001244298.1 | 6 - 7 | 927 | 63 | |
XM_015287091.2 | XP_015142577.1 | 6 - 7 | 964 | 63 | ||
GenBank mRNA | BX932917.2 | - | 6 - 7 | 927 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
