This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07160529_g1
See other POLR3H GE Assays ›
Gene Symbol:
POLR3H
Gene Name:
RNA polymerase III subunit H
Gene Aliases:
-
Chromosome Location:
Chr.1: 49601069 - 49607431 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07160529_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277374.1 NP_001264303.1 5 - 6 422 104
XM_015285093.2 XP_015140579.1 5 - 6 427 104
GenBank mRNA BX935642.2 - 5 - 6 410 104
BX936031.1 - 4 - 5 390 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9j6kv:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0