This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07160540_m1
See other VTI1A GE Assays ›
Gene Symbol:
VTI1A
Gene Name:
vesicle transport through interaction with t-SNAREs 1A
Gene Aliases:
-
Chromosome Location:
Chr.6: 27693934 - 27946083 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07160540_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VTI1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277400.1 NP_001264329.1 1 - 2 162 62
NM_001277401.1 NP_001264330.1 1 - 2 162 62
XM_015288701.2 XP_015144187.1 1 - 2 442 62
XM_015288702.2 XP_015144188.1 1 - 2 442 62
XM_015288703.2 XP_015144189.2 1 - 2 170 62
XM_015288704.2 XP_015144190.1 1 - 2 442 62
XM_015288705.2 XP_015144191.1 1 - 2 442 62
XM_015288706.2 XP_015144192.1 1 - 2 442 62
XM_015288707.2 XP_015144193.1 1 - 2 442 62
XM_025151647.1 XP_025007415.1 1 - 2 442 62
XM_025151648.1 XP_025007416.1 1 - 2 442 62
XM_025151649.1 XP_025007417.1 1 - 2 442 62
GenBank mRNA BM491512.1 - 1 - 2 161 62
CO421979.1 - 1 - 2 143 62
CV858513.1 - 1 - 2 145 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0