This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07160631_m1
See other MGST3 GE Assays ›
Gene Symbol:
MGST3
Gene Name:
microsomal glutathione S-transferase 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 5849191 - 5858525 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07160631_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277533.1 NP_001264462.1 4 - 5 562 90
NM_001277537.1 NP_001264466.1 4 - 5 309 90
GenBank mRNA BU355153.1 - 4 - 5 309 90
CN383177.1 - 4 - 5 359 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline