This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07161027_g1
See other ESD GE Assays ›
Gene Symbol:
ESD
Gene Name:
esterase D
Gene Aliases:
-
Chromosome Location:
Chr.1: 169645532 - 169655054 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07161027_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277791.1 NP_001264720.1 3 - 4 340 83
NM_001277792.1 NP_001264721.1 3 - 4 215 83
NM_001277793.1 NP_001264722.1 3 - 4 198 83
XM_015275581.2 XP_015131067.1 3 - 4 247 83
XM_025150364.1 XP_025006132.1 3 - 4 343 83
XM_025150365.1 XP_025006133.1 3 - 4 396 83
XM_025150373.1 XP_025006141.1 3 - 4 216 83
GenBank mRNA BM427320.1 - 3 - 4 325 83
BU340762.1 - 3 - 4 215 83
BX930968.2 - 3 - 4 198 83
BX934861.1 - 3 - 4 200 83
CR385455.1 - 3 - 4 215 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0