This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07161071_g1
See other ATP5G3 GE Assays ›
Gene Symbol:
ATP5G3
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Gene Aliases:
ATP5MC3
Chromosome Location:
Chr.7: 16648520 - 16651688 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07161071_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5G3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277855.1 NP_001264784.1 2 - 3 255 78
NM_001277856.1 NP_001264785.1 2 - 3 255 78
NR_102686.1 2 - 3 255 78
GenBank mRNA BU404648.1 - 2 - 3 255 78
BU468804.1 - 2 - 3 178 78
CN220040.1 - 2 - 3 113 78
CN225005.1 - 2 - 3 129 78
CO635700.1 - 2 - 3 156 78
CR385453.1 - 2 - 3 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0