This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07161355_g1
See other RPL34 GE Assays ›
Gene Symbol:
RPL34
Gene Name:
ribosomal protein L34
Gene Aliases:
-
Chromosome Location:
Chr.4: 37708385 - 37711694 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07161355_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001281500.1 NP_001268429.1 2 - 3 98 92
XM_015276204.2 XP_015131690.1 2 - 3 186 92
XM_015276205.1 XP_015131691.1 2 - 3 97 92
GenBank mRNA BX259043.3 - 1 - 2 65 92
BX931507.1 - 1 - 2 556 92
BX932642.1 - 1 - 2 569 92
BX950575.1 - 2 - 3 334 92
CF251027.1 - 2 - 3 100 92
DR428690.1 - 1 - 2 85 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline