This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07162510_g1
See other PGGHG GE Assays ›
Gene Symbol:
PGGHG
Gene Name:
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases:
ATHL1
Chromosome Location:
Chr.5: 1823910 - 1835830 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07162510_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGGHG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318401.1 NP_001305330.1 13 - 14 2068 60
XM_015286261.2 XP_015141747.1 13 - 14 2148 60
XM_015286263.2 XP_015141749.1 13 - 14 2344 60
XM_015286264.2 XP_015141750.1 13 - 14 3418 60
XM_015286265.2 XP_015141751.1 13 - 14 2211 60
XM_025150650.1 XP_025006418.1 13 - 14 2558 60
XM_025150651.1 XP_025006419.1 12 - 13 2537 60
GenBank mRNA CR405906.1 - 4 - 5 554 60
LC011569.1 - 13 - 14 2044 60
LC011570.1 - 13 - 14 2068 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
hydrolase activity, hydrolyzing O-glycosyl compounds
protein-glucosylgalactosylhydroxylysine glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0