This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07163788_g1
See other ATP5G2 GE Assays ›
Gene Symbol:
ATP5G2 ATF7
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
activating transcription factor 7
Gene Aliases:
ATP5MC2
-
Chromosome Location:
Chr.33: 7481190 - 7483149 on Build GRCg6a
Chromosome Location:
Chr.33: 7433931 - 7483103 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
-
Assay ID Gg07163788_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5G2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015300350.2 XP_015155836.1 2 - 3 163 82
GenBank mRNA BX929366.2 - 1 - 2 99 82
BX931315.2 - 2 - 3 114 82
BX934992.1 - 101 82
Gene Symbol: ATF7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_025145448.1 XP_025001216.1 2 - 3 117 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_025145448.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0