This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07163789_g1
See other ATP5G2 GE Assays ›
Gene Symbol:
ATP5G2
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Gene Aliases:
ATP5MC2
Chromosome Location:
Chr.33: 7481190 - 7483149 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07163789_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5G2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015300350.2 XP_015155836.1 3 - 4 235 93
GenBank mRNA BX929366.2 - 2 - 3 171 93
BX931315.2 - 3 - 4 186 93
BX934992.1 - 173 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0