This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07164749_m1
See other CHTOP GE Assays ›
Gene Symbol:
CHTOP
Gene Name:
chromatin target of PRMT1
Gene Aliases:
C1orf77, C25H1orf77
Chromosome Location:
Chr.25: 3329538 - 3335937 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07164749_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHTOP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004948216.3 XP_004948273.1 2 - 3 274 64
XM_015298558.2 XP_015154044.1 2 - 3 278 64
XM_015298559.2 XP_015154045.1 2 - 3 285 64
XM_015298561.2 XP_015154047.1 2 - 3 318 64
XM_025143486.1 XP_024999254.1 2 - 3 285 64
XM_025143487.1 XP_024999255.1 2 - 3 295 64
XM_025143488.1 XP_024999256.1 2 - 3 273 64
XM_424013.6 XP_424013.3 2 - 3 272 64
GenBank mRNA CR353792.1 - 2 - 3 216 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline