This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07166068_s1
See other GPR1 GE Assays ›
Gene Symbol:
GPR1
Gene Name:
G protein-coupled receptor 1
Gene Aliases:
GPR1-va, GPR1-vb, GPR1-vc
Chromosome Location:
Chr.7: 12546633 - 12571393 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07166068_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004942654.3 XP_004942711.1 2 3924 118
XM_015289516.2 XP_015145002.1 2 2351 118
XM_025152432.1 XP_025008200.1 2 2222 118
XM_025152433.1 XP_025008201.1 2 2399 118
GenBank mRNA CR386532.1 - 1 126 118
KX156840.1 - 2 2222 118
KX156841.1 - 3 1727 118
KX156842.1 - 3 1690 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline