This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07167224_s1
See other ATF3 GE Assays ›
Gene Symbol:
ATF3
Gene Name:
activating transcription factor 3
Gene Aliases:
-
Chromosome Location:
Chr.3: 21943596 - 21967757 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07167224_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015283874.2 XP_015139360.1 4 1689 99
XM_015283877.2 XP_015139363.1 4 1693 99
XM_015283878.2 XP_015139364.1 4 1599 99
XM_015283879.1 XP_015139365.1 4 2806 99
XM_025149330.1 XP_025005098.1 4 1719 99
XM_025149331.1 XP_025005099.1 5 1761 99
XM_025149332.1 XP_025005100.1 5 1857 99
GenBank mRNA CR389106.1 - 1 414 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline