This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07169154_m1
See other VOPP1 GE Assays ›
Gene Symbol:
VOPP1
Gene Name:
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 51667730 - 51738595 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07169154_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VOPP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001235487.5 XP_001235488.1 1 - 2 407 63
XM_004939605.3 XP_004939662.1 1 - 2 458 63
XM_025147590.1 XP_025003358.1 1 - 2 298 63
GenBank mRNA BX929978.1 - 1 - 2 77 63
BX950508.2 - 1 - 2 148 63
CR407361.1 - 1 - 2 388 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_025147590.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline