This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07169292_g1
See other ATP5O GE Assays ›
Gene Symbol:
ATP5O
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases:
-
Chromosome Location:
Chr.1: 106924124 - 106927541 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07169292_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5O
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_416717.6 XP_416717.4 6 - 7 649 115
GenBank mRNA BX930173.1 - 6 - 7 541 115
BX930898.1 - 6 - 7 559 115
BX931279.1 - 1 - 2 859 115
BX932744.1 - 4 - 5 298 115
BX950594.1 - 6 - 7 548 115
CR407464.1 - 3 - 4 536 115
CR733382.1 - 2 - 3 719 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0