This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07170399_m1
See other YPEL1 GE Assays ›
Gene Symbol:
YPEL1
Gene Name:
yippee like 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 847763 - 862843 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07170399_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YPEL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015275692.2 XP_015131178.1 3 - 4 469 101
XM_415068.6 XP_415068.1 3 - 4 621 101
GenBank mRNA CR523261.1 - 3 - 4 542 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline