This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07171393_g1
See other LOC428277 GE Assays ›
Gene Symbol:
LOC428277
Gene Name:
parathyroid hormone/parathyroid hormone-related peptide receptor-like
Gene Aliases:
PTH3R, PTHRL
Chromosome Location:
Chr.27: 5241219 - 5250731 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07171393_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC428277
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_425837.6 XP_425837.4 5 - 6 3123 92
GenBank mRNA EU250015.1 - 5 - 6 523 92
FR746110.1 - 5 - 6 499 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline