This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07171542_m1
See other MSRA GE Assays ›
Gene Symbol:
MSRA
Gene Name:
methionine sulfoxide reductase A
Gene Aliases:
-
Chromosome Location:
Chr.3: 106426060 - 106687405 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07171542_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004935891.3 XP_004935948.1 1 - 2 779 93
XM_015285182.2 XP_015140668.1 1 - 2 133 93
XM_025149164.1 XP_025004932.1 1 - 2 779 93
XM_420035.6 XP_420035.3 1 - 2 159 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fxmhk:80/100.66.79.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0