This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07174908_m1
See other GDE1 GE Assays ›
Gene Symbol:
GDE1
Gene Name:
glycerophosphodiester phosphodiesterase 1
Gene Aliases:
-
Chromosome Location:
Chr.14: 8912439 - 8932362 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07174908_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015294545.2 XP_015150031.1 2 - 3 579 86
XM_015294546.2 XP_015150032.1 2 - 3 579 86
XM_015294547.2 XP_015150033.1 2 - 3 579 86
XM_015294548.2 XP_015150034.1 2 - 3 579 86
XM_025155327.1 XP_025011095.1 2 - 3 579 86
XM_025155328.1 XP_025011096.1 2 - 3 579 86
XM_025155329.1 XP_025011097.1 2 - 3 579 86
XM_025155330.1 XP_025011098.1 2 - 3 387 86
XM_025155331.1 XP_025011099.1 2 - 3 309 86
GenBank mRNA BX930663.1 - 2 - 3 421 86
BX932792.2 - 2 - 3 431 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

phosphodiesterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline