This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07178503_s1
See other GLCE GE Assays ›
Gene Symbol:
GLCE
Gene Name:
glucuronic acid epimerase
Gene Aliases:
-
Chromosome Location:
Chr.10: 19727395 - 19763603 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07178503_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLCE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003641864.4 XP_003641912.1 4 3178 102
XM_004943933.3 XP_004943990.1 4 3189 102
XM_015292209.2 XP_015147695.1 4 3140 102
XM_015292210.2 XP_015147696.1 7 3793 102
XM_015292212.1 XP_015147698.1 6 3427 102
XM_015292214.2 XP_015147700.1 5 3503 102
XM_025153877.1 XP_025009645.1 6 3604 102
XM_025153879.1 XP_025009647.1 6 3541 102
XM_025153880.1 XP_025009648.1 5 3238 102
XM_025153881.1 XP_025009649.1 4 2811 102
XM_025153882.1 XP_025009650.1 4 2793 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline