This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07180582_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.Z: 67623582 - 67708615 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07180582_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004949537.3 XP_004949594.1 2 - 3 308 102
XM_004949539.3 XP_004949596.1 2 - 3 309 102
XM_004949541.3 XP_004949598.1 2 - 3 309 102
XM_004949544.3 XP_004949601.1 3 - 4 309 102
XM_015280735.2 XP_015136221.1 4 - 5 599 102
XM_015280736.2 XP_015136222.1 4 - 5 328 102
XM_015280739.2 XP_015136225.1 3 - 4 252 102
XM_015280740.1 XP_015136226.1 2 - 3 141 102
XM_025145029.1 XP_025000797.1 2 - 3 309 102
XM_025145031.1 XP_025000799.1 4 - 5 601 102
GenBank mRNA BX931268.1 - 2 - 3 149 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0