This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07184714_m1
See other COMMD1 GE Assays ›
Gene Symbol:
COMMD1
Gene Name:
copper metabolism domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 9595603 - 9666304 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07184714_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COMMD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001231775.5 XP_001231776.1 2 - 3 498 165
XM_004940200.3 XP_004940257.1 4 - 5 678 165
XM_015278114.2 XP_015133600.1 4 - 5 670 165
XM_025149390.1 XP_025005158.1 2 - 3 360 165
XM_025149391.1 XP_025005159.1 2 - 3 323 165
XM_025149392.1 XP_025005160.1 3 - 4 663 165
GenBank mRNA BX932712.2 - 2 - 3 484 165

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0