This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07184785_g1
See other PPIL1 GE Assays ›
Gene Symbol:
PPIL1
Gene Name:
peptidylprolyl isomerase like 1
Gene Aliases:
-
Chromosome Location:
Chr.26: 1188704 - 1190460 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07184785_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003643916.4 XP_003643964.1 268 94
XM_004950979.3 XP_004951036.1 262 94
XM_004950980.3 XP_004951037.1 302 94
XM_004950981.3 XP_004951038.1 266 94
XM_015298786.1 XP_015154272.1 300 94
GenBank mRNA BX932874.1 - 218 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0