This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07192511_m1
See other CALHM1 GE Assays ›
Gene Symbol:
CALHM1 CALHM2
Gene Name:
calcium homeostasis modulator 1
calcium homeostasis modulator 2
Gene Aliases:
FAM26C
FAM26B
Chromosome Location:
Chr.6: 24593725 - 24600892 on Build GRCg6a
Chromosome Location:
Chr.6: 24590712 - 24595374 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
-
Assay ID Gg07192511_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CALHM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015288733.2 XP_015144219.1 2 - 3 4912 58
Gene Symbol: CALHM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015288735.2 XP_015144221.1 1 - 2 206 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015288735.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0