This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07195578_m1
See other MOB3B GE Assays ›
Gene Symbol:
MOB3B
Gene Name:
MOB kinase activator 3B
Gene Aliases:
MOBKL2B
Chromosome Location:
Chr.Z: 68817124 - 68903773 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07195578_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004949585.3 XP_004949642.1 3 - 4 1003 75
XM_004949588.3 XP_004949645.1 3 - 4 1404 75
XM_004949590.3 XP_004949647.1 3 - 4 1017 75
XM_004949591.3 XP_004949648.1 2 - 3 916 75
XM_015280718.2 XP_015136204.1 2 - 3 1333 75
XM_025144871.1 XP_025000639.1 4 - 5 1096 75
XM_025144872.1 XP_025000640.1 4 - 5 1104 75
XM_025144873.1 XP_025000641.1 4 - 5 1111 75
XM_025144874.1 XP_025000642.1 4 - 5 1483 75
XM_025144875.1 XP_025000643.1 4 - 5 1009 75
XM_025144876.1 XP_025000644.1 3 - 4 923 75
XM_025144877.1 XP_025000645.1 4 - 5 1104 75
XM_025144878.1 XP_025000646.1 3 - 4 995 75
XM_025144879.1 XP_025000647.1 4 - 5 1491 75
XM_025144880.1 XP_025000648.1 3 - 4 1412 75
XM_025144881.1 XP_025000649.1 4 - 5 1082 75
XM_025144882.1 XP_025000650.1 3 - 4 1025 75
XM_025144883.1 XP_025000651.1 2 - 3 924 75
XM_025144884.1 XP_025000652.1 2 - 3 2222 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0