This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07198465_m1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 61566796 - 62027465 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07198465_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003640755.4 XP_003640803.1 3 - 4 2596 83
XM_015275961.2 XP_015131447.1 5 - 6 2579 83
XM_015275962.2 XP_015131448.1 5 - 6 2559 83
XM_015275963.2 XP_015131449.1 5 - 6 2677 83
XM_015275965.2 XP_015131451.1 6 - 7 2708 83
XM_015275966.2 XP_015131452.1 4 - 5 2573 83
XM_015275967.2 XP_015131453.1 3 - 4 2222 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline