This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07198473_m1
See other GCNT1 GE Assays ›
Gene Symbol:
GCNT1
Gene Name:
glucosaminyl (N-acetyl) transferase 1
Gene Aliases:
-
Chromosome Location:
Chr.Z: 37363170 - 37376183 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07198473_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003643022.4 XP_003643070.1 2 - 3 170 85
XM_004949175.3 XP_004949232.1 2 - 3 285 85
XM_015280355.2 XP_015135841.1 2 - 3 289 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_003643022.4, XM_004949175.3, XM_015280355.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0