This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07198625_g1
See other TP73 GE Assays ›
Gene Symbol:
TP73
Gene Name:
tumor protein p73
Gene Aliases:
-
Chromosome Location:
Chr.21: 946406 - 979523 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07198625_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TP73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004947368.3 XP_004947425.1 5 - 6 806 85
XM_025142621.1 XP_024998389.1 7 - 8 988 85
XM_025142622.1 XP_024998390.1 7 - 8 1224 85
XM_025142623.1 XP_024998391.1 8 - 9 1166 85
XM_025142624.1 XP_024998392.1 8 - 9 1111 85
XM_025142625.1 XP_024998393.1 8 - 9 1140 85
XM_025142626.1 XP_024998394.1 7 - 8 1051 85
XM_025142627.1 XP_024998395.1 7 - 8 1308 85
XM_025142628.1 XP_024998396.1 6 - 7 792 85
XM_025142629.1 XP_024998397.1 7 - 8 988 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

P53-like transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0