This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07211900_s1
See other GJB5 GE Assays ›
Gene Symbol:
GJB5
Gene Name:
gap junction protein beta 5
Gene Aliases:
-
Chromosome Location:
Chr.23: 4606221 - 4611466 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07211900_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004947782.3 XP_004947839.1 1 2671 70
XM_015297824.2 XP_015153310.1 1 2706 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004947782.3, XM_015297824.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline