This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm03760494_m1
See other BZIP68 GE Assays ›
Gene Symbol:
BZIP68
Gene Name:
bZIP transcription factor bZIP68
Gene Aliases:
-
Chromosome Location:
Chr.2: 18951005 - 18954047 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm03760494_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BZIP68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001250115.2 NP_001237044.1 1 - 2 430 60
GenBank mRNA AK245520.1 - 1 - 2 412 60
DQ787040.1 - 1 - 2 434 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Glycine max Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0