Assay Details
| Gene Symbol: LOC100793134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252950.1 | NP_001239879.1 | 1 - 2 | 537 | 63 | |
| GenBank mRNA | AK286139.1 | - | 1 - 2 | 487 | 63 | |
| BT096853.1 | - | 1 - 2 | 537 | 63 | ||
More Information
Set Membership: |
|

LOC100793134
Genomic Map