This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg00392011_m1
See other GLCE GE Assays ›
Gene Symbol:
Glce
Gene Name:
glucuronic acid epimerase
Gene Aliases:
GW7_09956
Chromosome Location:
Chr.Un|NW_004624781: 4135829 - 4228948 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg00392011_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855572.2 XP_004855629.1 4 - 5 889 68
XM_004855574.2 XP_004855631.1 3 - 4 818 68
XM_004855575.2 XP_004855632.1 2 - 3 717 68
XM_013076231.2 XP_012931685.1 5 - 6 977 68
XM_013076232.2 XP_012931686.1 4 - 5 877 68
XM_021237004.1 XP_021092663.1 3 - 4 815 68
XM_021237006.1 XP_021092665.1 4 - 5 899 68
XM_021237007.1 XP_021092666.1 4 - 5 895 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline