This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg00504689_g1
See other RBM39 GE Assays ›
Gene Symbol:
Rbm39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
GW7_21268
Chromosome Location:
Chr.Un|NW_004624842: 3698631 - 3733221 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg00504689_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867860.3 XP_004867917.1 4 - 5 605 58
XM_004867861.3 XP_004867918.1 4 - 5 605 58
XM_021246832.1 XP_021102491.1 4 - 5 606 58
XM_021246833.1 XP_021102492.1 5 - 6 1203 58
XM_021246834.1 XP_021102493.1 2 - 3 1685 58
XM_021246835.1 XP_021102494.1 6 - 7 750 58
XM_021246836.1 XP_021102495.1 6 - 7 1275 58
XM_021246837.1 XP_021102496.1 4 - 5 382 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021246833.1, XM_021246834.1, XM_021246835.1, XM_021246836.1, XM_021246837.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0