This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg01402200_m1
See other CADM2 GE Assays ›
Gene Symbol:
Cadm2
Gene Name:
cell adhesion molecule 2
Gene Aliases:
GW7_06803
Chromosome Location:
Chr.Un|NW_004624789: 1244772 - 2315267 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg01402200_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004857903.3 XP_004857960.1 5 - 6 702 80
XM_021238782.1 XP_021094441.1 6 - 7 781 80
XM_021238783.1 XP_021094442.1 5 - 6 632 80
XM_021238784.1 XP_021094443.1 5 - 6 758 80
XM_021238785.1 XP_021094444.1 6 - 7 781 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0